Learn More
Sino Biological Human IL-1RAcP / IL-1R3 Protein (Fc Tag) 100ug
Supplier:  Sino Biological 10121H02H100
Encompass_Preferred
A DNA sequence encoding the human IL1RAP (NP_002173.1) (Met1-Glu359) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-4892