Learn More
ACROBiosystems Recombinant Protein;Human IL-1 RII / CD121b Protein, Fc Tag;HEK293;1MG;IL2,CD121b,IL1RB,IL1R2,IL-1R2,CDw121b,IL-1R-2,IL-1RT-2,IL-1RT2,IL-1RB,IL1R,IL-1R,IL-1 RII
Catalog No. 501657976
Encompass_Preferred
Recombinant Protein;Human IL-1 RII / CD121b Protein, Fc Tag;1MG;Human IL-1 RII, Fc Tag (IL2-H4256) is expressed from human 293 cells (HEK293). It contains AA Phe 14 - Glu 343 (Accession # NP_004624.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64 kDa. The protein migrates as 68-74 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-1 RII, Fc Tag (Cat. No. IL2-H4256) at 10 μg/mL (100 µL/well),can bind Biotinylated Human IL-1 alpha (Cat. No. ILA-H8213) with a linear range of 0.05-1 μg/mL (QC tested).
Catalog No. 50-165-7976 Supplier ACROBiosystems Supplier No. IL2H42561MG
Recombinant Protein;Human IL-1 RII / CD121b Protein, Fc Tag;1MG;Human IL-1 RII, Fc Tag (IL2-H4256) is expressed from human 293 cells (HEK293). It contains AA Phe 14 - Glu 343 (Accession # NP_004624.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64 kDa. The protein migrates as 68-74 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-1 RII, Fc Tag (Cat. No. IL2-H4256) at 10 μg/mL (100 µL/well),can bind Biotinylated Human IL-1 alpha (Cat. No. ILA-H8213) with a linear range of 0.05-1 μg/mL (QC tested).