Learn More
ACROBiosystems Recombinant Protein;Human IL-1 RII / CD121b Protein;HEK293;100UG;IL2,CD121b,IL1RB,IL1R2,IL-1R2,CDw121b,IL-1R-2,IL-1RT-2,IL-1RT2,IL-1RB,IL1R,IL-1R,IL-1 RII
Supplier:  ACROBiosystems IL2H4226100UG
Encompass_Preferred
Recombinant Protein;Human IL-1 RII / CD121b Protein;100UG;Human IL-1 RII, His Tag (IL2-H4226) is expressed from human 293 cells (HEK293). It contains AA Thr 14 - Glu 343 (Accession # NP_004624.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 38.6 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7854