Learn More
ACROBiosystems Recombinant Protein;Human IL-1 Rrp2 / IL-1 R6 Protein, Fc Tag;HEK293;100UG;IL2,IL-1 R6,IL-1 Rrp2,IL1RL2,IL-1RL2,IL1Rrp2,IL-1Rrp2,IL1RRP2,IL-36R,IL-36 receptor
Catalog No. 501657640
Encompass_Preferred
Recombinant Protein;Human IL-1 Rrp2 / IL-1 R6 Protein, Fc Tag;100UG;Human IL-1 Rrp2, Fc Tag (IL2-H5254) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 335 (Accession # Q9HB29-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 62.4 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Recombinant Human IL-36Ra / IL-1F5 at 2 μg/mL (100 μL/well) can bind Human IL-1 Rrp2, Fc Tag (Cat. No. IL2-H5254) with a linear range of 0.156-5 μg/mL (QC tested).
Catalog No. 50-165-7640 Supplier ACROBiosystems Supplier No. IL2H5254100UG
Recombinant Protein;Human IL-1 Rrp2 / IL-1 R6 Protein, Fc Tag;100UG;Human IL-1 Rrp2, Fc Tag (IL2-H5254) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 335 (Accession # Q9HB29-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 62.4 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Recombinant Human IL-36Ra / IL-1F5 at 2 μg/mL (100 μL/well) can bind Human IL-1 Rrp2, Fc Tag (Cat. No. IL2-H5254) with a linear range of 0.156-5 μg/mL (QC tested).