Learn More
Sino Biological IL2 Antibody (APC), Mouse MAb 100Tests
Supplier:  Sino Biological 11848MM03A100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL2 (rh IL2; Catalog#11848-HNAE; P60568-1; Ala21-Thr153) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-224-6379