Learn More
Sino Biological IL2 Antibody (FITC), Rabbit MAb 25Tests
Supplier:  Sino Biological 51061R300F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL2 (rM IL2; Catalog#51061-MNAE; P04351; Ala21-Gln169) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-224-6533