Learn More
Sino Biological IL-2 Antibody (HRP), Rabbit PAb 100ug
Supplier:  Sino Biological 11848RP01H100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL-2 (rh IL-2; Catalog#11848-HNAE; P60568-1; Ala21-Thr153). Total IgG was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-224-7537