Learn More
Sino Biological Recombinant Anti-IL-21 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50137R012100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-21 (rM IL-21; Catalog#50137-MNAE; Q9ES17; Pro25-Ser146).
Catalog No. 50-228-2714