Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP539410UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: Untagged. Host: E. coli. Species Reactivity: Human. BioActivity: Measured by mino cell proliferation. AA Sequence: MLKTLNLGSC VIATNLQEIR NGFSEIRGSV QAKDGNIDIR ILRRTESLQD TKPANRCCLL RHLLRLYLDR VFKNYQTPDH YTLRKISSLA NSFLTIKKDL RLCHAHMTCH CGEEAMKKYS QILSHFEKLE PQAAVVKALG ELDILLQWME ETE. Buffer: 10 mM sodium phosphate, pH 7.5. Purity: Greater than 95% as determined by SDS-PAGE. Endotoxin: Less than or equal to 1.00 EUs/ug as determined by the LAL method. Gene Symbol: IL21. Gene ID: 59067. Uniprot: Q9HBE4.
Catalog No. 50-188-5810