Learn More
Sino Biological Rat IL-21 Protein 20ug
Supplier:  Sino Biological 80200RNAE20
Encompass_Preferred
A DNA sequence encoding the rat IL21 (NP_001102413.1) (Pro25-Ser146) was expressed with an initial Met.
Catalog No. 50-211-5034