Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human IL-21 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;IL1,IL21,CVID11,IL-21,Za11
Catalog No. 501658382
Encompass_Preferred
Recombinant Protein;Biotinylated Human IL-21 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human IL-21, Fc Tag (IL1-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Ser 155 (Accession # Q9HBE4-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 43.9 kDa. The protein migrates as 36 kDa and 48-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human IL-21, Fc Tag (Cat. No. IL1-H82F7) at 2 μg/mL (100 μL/well) can bind Human IL-21 R, His Tag (Cat. No. ILR-H5226) with a linear range of 2-8 ng/mL (QC tested).
Catalog No. 50-165-8382 Supplier ACROBiosystems Supplier No. IL1H82F725UG
Recombinant Protein;Biotinylated Human IL-21 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human IL-21, Fc Tag (IL1-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Ser 155 (Accession # Q9HBE4-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 43.9 kDa. The protein migrates as 36 kDa and 48-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human IL-21, Fc Tag (Cat. No. IL1-H82F7) at 2 μg/mL (100 μL/well) can bind Human IL-21 R, His Tag (Cat. No. ILR-H5226) with a linear range of 2-8 ng/mL (QC tested).