Learn More
Sino Biological IL21R Protein 100ug (Sino Biological)
Supplier:  Sino Biological 10279MM17TF100
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus IL21R (XP_005591578.1) (Met1-Pro236) was expressed with the Fc region of human IgG1 at the C-terminus. Cynomolgus and Rhesus IL21R sequences are identical.
Catalog No. 50-237-4028