Learn More
ACROBiosystems Recombinant Protein;Human IL-21 R Protein, Fc Tag;HEK293;1MG;ILR,IL21R,CD360,NILR,IL-21R
Catalog No. 501658472
Encompass_Preferred
Recombinant Protein;Human IL-21 R Protein, Fc Tag;1MG;Human IL-21 R, Fc Tag (ILR-H5257) is expressed from human 293 cells (HEK293). It contains AA Cys 20 - Pro 236 (Accession # AAH04348).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.6 kDa. As a result of glycosylation, the protein migrates as 65-80 kDa under reducing (R) condition, and 130-160 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Biotinylated Human IL-21, Fc Tag (Cat. No. IL1-H82F7) at 2 μg/mL (100 μL/well) can bind Human IL-21 R, Fc Tag (Cat. No. ILR-H5257) with a linear range of 2-31 ng/mL (QC tested).
Catalog No. 50-165-8472 Supplier ACROBiosystems Supplier No. ILRH52571MG
Recombinant Protein;Human IL-21 R Protein, Fc Tag;1MG;Human IL-21 R, Fc Tag (ILR-H5257) is expressed from human 293 cells (HEK293). It contains AA Cys 20 - Pro 236 (Accession # AAH04348).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.6 kDa. As a result of glycosylation, the protein migrates as 65-80 kDa under reducing (R) condition, and 130-160 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Biotinylated Human IL-21, Fc Tag (Cat. No. IL1-H82F7) at 2 μg/mL (100 μL/well) can bind Human IL-21 R, Fc Tag (Cat. No. ILR-H5257) with a linear range of 2-31 ng/mL (QC tested).