Learn More
ACROBiosystems Recombinant Protein;Human IL-21 R Protein;HEK293;200UG;ILR,IL21R,CD360,NILR,IL-21R
Catalog No. 501658469
Encompass_Preferred
Recombinant Protein;Human IL-21 R Protein;200UG;Human IL-21 R, His Tag (ILR-H5226) is expressed from human 293 cells (HEK293). It contains AA Cys 20 - Pro 236 (Accession # AAH04348).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.8 kDa. The protein migrates as 42-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human IL-21, Fc Tag (Cat. No. IL1-H82F7) at 2 μg/mL (100 μL/well) can bind Human IL-21 R, His Tag (Cat. No. ILR-H5226) with a linear range of 2-8 ng/mL (QC tested).
Catalog No. 50-165-8469 Supplier ACROBiosystems Supplier No. ILRH5226200UG
Recombinant Protein;Human IL-21 R Protein;200UG;Human IL-21 R, His Tag (ILR-H5226) is expressed from human 293 cells (HEK293). It contains AA Cys 20 - Pro 236 (Accession # AAH04348).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.8 kDa. The protein migrates as 42-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human IL-21, Fc Tag (Cat. No. IL1-H82F7) at 2 μg/mL (100 μL/well) can bind Human IL-21 R, His Tag (Cat. No. ILR-H5226) with a linear range of 2-8 ng/mL (QC tested).