Learn More
Aviva Systems Biology Corp IL22 Antibody - C-terminal region (ARP49523_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49523_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL22. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human IL22. Alternate names: IL-21, IL-22, IL-D110, IL-TIF, IL21, ILTIF, MGC79382, MGC79384, TIFIL-23, TIFa, zcyto18ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-60101