Learn More
ACROBiosystems Recombinant Protein;Human IL-22 Protein, His Tag;HEK293;1MG;IL2,IL22,IL-22,ILTIF,IL-TIF,ZCYTO18
Catalog No. 501657457
Encompass_Preferred
Recombinant Protein;Human IL-22 Protein, His Tag;1MG;Human IL-22, His Tag (IL2-H524a) is expressed from human 293 cells (HEK293). It contains AA Ala 34 - Ile 179 (Accession # Q9GZX6-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 18.6 kDa. The protein migrates as 28-34 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-22, His Tag (Cat. No. IL2-H524a) at 5 μg/mL (100 μL/well) can bind Anti-Human IL-22 MAb with a linear range of 0.5-16 ng/mL (QC tested).
Catalog No. 50-165-7457 Supplier ACROBiosystems Supplier No. IL2H524A1MG
Recombinant Protein;Human IL-22 Protein, His Tag;1MG;Human IL-22, His Tag (IL2-H524a) is expressed from human 293 cells (HEK293). It contains AA Ala 34 - Ile 179 (Accession # Q9GZX6-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 18.6 kDa. The protein migrates as 28-34 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-22, His Tag (Cat. No. IL2-H524a) at 5 μg/mL (100 μL/well) can bind Anti-Human IL-22 MAb with a linear range of 0.5-16 ng/mL (QC tested).