Learn More
Aviva Systems Biology Corp IL22RA1 Antibody - middle region (ARP49651_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49651_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL22RA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IL22RA1. Alternate names: CRF2-9, IL22R, IL22R1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60137