Learn More
Sino Biological IL22RA2 / IL22BP Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11025MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL22RA2 / IL22BP (rh IL22BP; Catalog#11025-H08H; NP_851826.1; Met 1-Pro 231). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-7726