Learn More
Sino Biological Human IL23R / IL23 Receptor Protein (His Tag) 100ug
Supplier:  Sino Biological 13840H08H100
Encompass_Preferred
A DNA sequence encoding the human IL23R (AAM44229.1) (Met1-Asp353) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-4524