Learn More
ACROBiosystems ACROBiosystems Mouse IL-23 alpha&IL-12 beta Heterodimer Protein, His Tag&Tag Free
Catalog No. 501957349
Encompass_Preferred
Recombinant Protein;1MG;Mouse IL-23A&IL-12B Heterodimer Protein, His Tag&Tag Free (ILB-M52W7) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Ala 196 (IL23A) & Met 23 - Ser 335 (IL12B) (Accession # Q9EQ14-1 (IL23A) & P43432-1 (IL12B)).;Mouse IL-23A&IL-12B Heterodimer Protein, His Tag&Tag Free, produced by co-expression of IL-23A and IL-12B, has a calculated MW of 20.5 kDa (IL-23A) and 35.8 kDa (IL-12B). Subunit IL-23A is fused with a polyhistidine tag at the N-terminus and subunit IL-12B contains no tag. The reducing (R) protein migrates as 21 kDa (IL-23A) and 45-50 kDa (IL-12B) respectively due to glycosylation.;Immobilized Human IL-23 R, Fc Tag (Cat. No. ILR-H5254) at 10 μg/mL (100 μL/well) can bind Mouse IL-23A&IL-12B Heterodimer Protein, His Tag&Tag Free (Cat. No. ILB-M52W7)
Catalog No. 50-195-7349 Supplier ACROBiosystems Supplier No. ILBM52W71MG
Add to Cart
Add to Cart
Request a Quote
Recombinant Protein;1MG;Mouse IL-23A&IL-12B Heterodimer Protein, His Tag&Tag Free (ILB-M52W7) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Ala 196 (IL23A) & Met 23 - Ser 335 (IL12B) (Accession # Q9EQ14-1 (IL23A) & P43432-1 (IL12B)).;Mouse IL-23A&IL-12B Heterodimer Protein, His Tag&Tag Free, produced by co-expression of IL-23A and IL-12B, has a calculated MW of 20.5 kDa (IL-23A) and 35.8 kDa (IL-12B). Subunit IL-23A is fused with a polyhistidine tag at the N-terminus and subunit IL-12B contains no tag. The reducing (R) protein migrates as 21 kDa (IL-23A) and 45-50 kDa (IL-12B) respectively due to glycosylation.;Immobilized Human IL-23 R, Fc Tag (Cat. No. ILR-H5254) at 10 μg/mL (100 μL/well) can bind Mouse IL-23A&IL-12B Heterodimer Protein, His Tag&Tag Free (Cat. No. ILB-M52W7)