Learn More
ACROBiosystems ACROBiosystems Human IL-23 alpha&IL-12 beta Heterodimer Protein, His Tag&Tag Free
Catalog No. 501957315
Encompass_Preferred
Recombinant Protein;50UG;Human IL-23A&IL-12B Heterodimer Protein, His Tag&Tag Free (ILB-H52W5) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Pro 189 (IL23A) & Ile 23 - Ser 328 (IL12B) (Accession # Q9NPF7-1 (IL23A) & P29460-1 (IL12B)).;Human IL-23A&IL-12B Heterodimer Protein, His Tag&Tag Free, produced by co-expression of IL-23 alpha and IL-12 beta, has a calculated MW of 19.5 kDa (IL-23 alpha) and 34.7 kDa (IL-12 beta). Subunit IL-23 alpha is fused with a polyhistidine tag at the N-terminus and subunit IL-12 beta contains no tag. The reducing (R) protein migrates as 20 kDa (IL-23 alpha) and 37-45 kDa (IL-12 beta) respectively due to glycosylation.;Immobilized Human IL-23A&IL-12B Heterodimer Protein, His Tag&Tag Free (Cat. No. ILB-H52W5) at 2 μg/mL (100 μL/well) can bind Anti IL23A&IL12B P40 domain, Human IgG1 with a linear range of 0.05-3 ng/mL (QC tested).;Immobilized Human IL-23 R, Fc Tag (Cat. No. ILR-H5254) at 10 μg/mL (100 μL/well) can bind Human IL-23A&IL-12
Catalog No. 50-195-7315 Supplier ACROBiosystems Supplier No. ILBH52W550UG
Recombinant Protein;50UG;Human IL-23A&IL-12B Heterodimer Protein, His Tag&Tag Free (ILB-H52W5) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Pro 189 (IL23A) & Ile 23 - Ser 328 (IL12B) (Accession # Q9NPF7-1 (IL23A) & P29460-1 (IL12B)).;Human IL-23A&IL-12B Heterodimer Protein, His Tag&Tag Free, produced by co-expression of IL-23 alpha and IL-12 beta, has a calculated MW of 19.5 kDa (IL-23 alpha) and 34.7 kDa (IL-12 beta). Subunit IL-23 alpha is fused with a polyhistidine tag at the N-terminus and subunit IL-12 beta contains no tag. The reducing (R) protein migrates as 20 kDa (IL-23 alpha) and 37-45 kDa (IL-12 beta) respectively due to glycosylation.;Immobilized Human IL-23A&IL-12B Heterodimer Protein, His Tag&Tag Free (Cat. No. ILB-H52W5) at 2 μg/mL (100 μL/well) can bind Anti IL23A&IL12B P40 domain, Human IgG1 with a linear range of 0.05-3 ng/mL (QC tested).;Immobilized Human IL-23 R, Fc Tag (Cat. No. ILR-H5254) at 10 μg/mL (100 μL/well) can bind Human IL-23A&IL-12