Learn More
Sino Biological IL23R Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90123C02H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus IL23R [(Identical to the rhesus IL23R (F6XR82)] (Met1-Asp353) was expressed with the Fc region of human IgG1 at the C-terminus. Cynomolgus and Rhesus IL23R sequences are identical.
Catalog No. 50-237-1492