Learn More
Sino Biological IL23R Protein 1MG (Sino Biological)
Supplier: Sino Biological 90123C02H1
A DNA sequence encoding the cynomolgus / rhesus IL23R [(Identical to the rhesus IL23R (F6XR82)] (Met1-Asp353) was expressed with the Fc region of human IgG1 at the C-terminus. Cynomolgus and Rhesus IL23R sequences are identical.