Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP551210UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: Untagged. Host: E. coli. Species Reactivity: Rat. BioActivity: Rat IL25 induces IL-8 production from human PBMCs at concentrations greater than or equal to 100 ng/ml. AA Sequence: MCSHLPRCCP SKQQEFPEEW LKWNPAPVSP PEPLRHTHHP ESCRASKDGP LNSRAISPWS YELDRDLNRV PQDLYHARCL CPHCVSLQTG SHMDPMGNSV PLYHNQTVFY RRPCHGEQGA HGRYCLERRL YRVSLACVCV RPRMMA. Buffer: Sterile filtered (0.22 um) solution containing 0.1% TFA. Purity: Greater than 95% as determined by SDS-PAGE. Endotoxin: Less than or equal to 1.00 EUs/ug as determined by the LAL method. Gene Symbol: IL25. Gene ID: 501996. Uniprot: D3ZLB1.
Catalog No. 50-188-5874