Learn More
Aviva Systems Biology Corp IL26 Antibody - N-terminal region (ARP64771_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64771_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL26. It was validated on Western Blot. . Alternate names: AK155, IL-26ApplicationWB. Species reactivity: Dog, Human, Pig, Rabbit
Catalog No. 50-288-365