Learn More
Aviva Systems Biology Corp IL27 Antibody - C-terminal region (ARP60748_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60748_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL27. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human IL27. Alternate names: IL-27, IL-27A, IL27p28, IL30, MGC71873, p28, IL27AApplicationWB. Species reactivity: Human
Catalog No. 50-287-146