Learn More
Aviva Systems Biology Corp IL28B Antibody - C-terminal region (ARP63701_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63701_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL28B. It was validated on Western Blot. . Alternate names: IFNL3, IL-28B, IL28C, IL28BApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-056