Learn More
Sino Biological IL-28B/IFN-lambda-3 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11890MM18100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-28B/IFN-lambda-3 (rh IL-28B/IFN-lambda-3; Catalog#11890-H08H; NP_742151.2; Met1-Val196). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8454