Learn More
ACROBiosystems Recombinant Protein;Human IL-29 / IFN-lambda 1 Protein;HEK293;50UG;IL9,IL29,IFNL1,IFNL-1,IL-29,IFNλ1,IFNλ-1,Interleukin 29,Interleukin-29,IFN lambda 1,Interleukin29
Supplier:  ACROBiosystems IL9H422850UG
Encompass_Preferred
Recombinant Protein;Human IL-29 / IFN-lambda 1 Protein;50UG;Human IL-29, His Tag (IL9-H4228) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Thr 200 (Accession # NP_742152.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.1 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7676