Learn More
Sino Biological IL2/IL-2/Interleukin-2 Antibody (HRP), Rabbit PAb, Antigen Affinity Purified 100ug
Supplier:  Sino Biological 11848T16H100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL2/IL-2/Interleukin-2 (rh IL2/IL-2/Interleukin-2; Catalog#11848-HNAY1; NP_000577.2; Ala 21-Thr153, Leu 38 Met, Leu 39 Ser, Cys 145 Ala). IL2/IL-2/Interleukin-2 specific IgG was purified by Human IL2/IL-2/Interleukin-2 affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-224-8065