Learn More
Sino Biological IL2/IL-2/Interleukin-2 Antibody (PE), Mouse MAb 25Tests
Supplier:  Sino Biological 80075MM06P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat IL2/IL-2/Interleukin-2 (rR IL2/IL-2/Interleukin-2; Catalog#80075-RNAE; P17108; Ala21-Gln155) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-224-8230