Learn More
ACROBiosystems Recombinant Protein;Human IL-2 R alpha / CD25 Protein, Fc Tag;HEK293;1MG;ILA,IL2RA,CD25,p55,IL2-RA,IL-2-RA,IL-2 R alpha,IL-2Ra,IL-2,IL2
Catalog No. 501658776
Encompass_Preferred
Recombinant Protein;Human IL-2 R alpha / CD25 Protein, Fc Tag;1MG;Human IL-2 R alpha, Fc Tag (ILA-H5251) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession # NP_000408).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.4 Kda. The protein migrates as 60-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-2 R alpha, Fc Tag (Cat. No. ILA-H5251) at 2μg/mL (100 μL/well) can bind Human IL-2, Tag Free (Cat. No. IL2-H4216) with a linear range of 0.2-1.56 ng/mL (QC tested).
Catalog No. 50-165-8776 Supplier ACROBiosystems Supplier No. ILAH52511MG
Recombinant Protein;Human IL-2 R alpha / CD25 Protein, Fc Tag;1MG;Human IL-2 R alpha, Fc Tag (ILA-H5251) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession # NP_000408).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.4 Kda. The protein migrates as 60-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-2 R alpha, Fc Tag (Cat. No. ILA-H5251) at 2μg/mL (100 μL/well) can bind Human IL-2, Tag Free (Cat. No. IL2-H4216) with a linear range of 0.2-1.56 ng/mL (QC tested).