Learn More
ACROBiosystems Recombinant Protein;Human IL-2 R alpha / CD25 Protein;HEK293;1MG;ILA,IL2RA,CD25,p55,IL2-RA,IL-2-RA,IL-2 R alpha,IL-2Ra,IL-2,IL2
Supplier:  ACROBiosystems ILAH52H91MG
Encompass_Preferred
Recombinant Protein;Human IL-2 R alpha / CD25 Protein;1MG;Human IL-2 R alpha, His Tag (ILA-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession # NP_000408).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.6 kDa. The protein migrates as 34-46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-2, Tag Free (Cat. No. IL2-H4216) at 5μg/mL (100 μL/well) can bind Human IL-2 R alpha, His Tag (Cat. No. ILA-H52H9) with a linear range of 0.01-1.11 μg/mL (QC tested).
Catalog No. 50-165-8775