Learn More
Sino Biological Human IL2RG Protein (His Tag) 1mg
Supplier:  Sino Biological 10555H08H1
Encompass_Preferred
A DNA sequence encoding the human IL2RG (NP_000197.1) (Met1-Ala262) was expressed with a polyhistide tag at the C-terminus.
Catalog No. 50-211-6405