Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP547710UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: Untagged. Host: E. coli. Species Reactivity: Mouse. BioActivity: Mouse IL-31 / IL31 induces IL-8 production in human PBMCs which was detected via ELISA (typical ED50 is ? 25 ng/ml). AA Sequence: MTCSLSFGAP ISKEDLRTTI DLLKQESQDL YNNYSIKQAS GMSADESIQL PCFSLDREAL TNISVIIAHL EKVKVLSENT VDTSWVIRWL TNISCFNPLN LNISVPGNTD ESYDCKVFVL TVLKQFSNCM AELQAKDNTT C. Buffer: 10 mM sodium phosphate, pH 7.5. Purity: Greater than 95% as determined by SDS-PAGE. Endotoxin: Less than or equal to 1.00 EUs/ug as determined by the LAL method. Gene Symbol: IL31. Gene ID: 76399. Uniprot: Q6EAL8.
Catalog No. 50-188-5854