Learn More
Sino Biological IL-32 alpha / IL32A Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11064MM12100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-32 alpha / IL32A (rh IL-32 alpha / IL32A; Catalog#11064-H08H; NP_001012651.1; Met1-Lys131). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8015