Learn More
Sino Biological IL32 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 11064R034100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL32 (rh IL32; Catalog#11064-H08H; NP_001012651.1; Met1-Lys131).
Catalog No. 50-224-7804