Learn More
Aviva Systems Biology Corp IL33 Antibody - N-terminal region (ARP51170_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51170_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL33. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IL33. Alternate names: C9orf26, DKFZp586H0523, DVS27, NF-HEV, NFEHEV, RP11-575C20.2, IL1F11ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-60509