Learn More
AdipoGen IL-36alpha (aa 6-158) (human) (rec.) (untagged) (3 x 10 µg)
SDP
Supplier:  AdipoGen AG40B01653010
Encompass_Preferred
Protein. Human IL-36alpha (aa Lys6-Phe158) is untagged. Source: E. coli. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Purity: >95% (SDS-PAGE). IL-36alpha (IL-1F6), IL-36beta (IL-1F8) and IL-36gamma (IL-1F9) bind to IL-36R (IL-1Rrp2) and IL-1RAcP, activating similar intracellular signals as IL-1 and are inhibited by IL-36Ra. IL-36 family members induce the production of proinflammatory cytokines, including IL-12, IL-1beta, IL-6, TNF-alpha and IL-23 in BMDC and CD4 T cells. Skin and dendritic cells are targets of the IL-36 interleukins leading to a Th1 response. These cytokines may represent potential targets for immune-mediated inflammatory conditions or, alternatively, could be used as adjuvants in vaccination. Recently a novel role for IL-36alpha in the pathogenesis of intestinal inflammation has been reported.
Catalog No. 50-205-3258