Learn More
Sino Biological IL3/IL-3/Interleukin-3 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11858MM35100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL3/IL-3/Interleukin-3 (rh IL3/IL-3/Interleukin-3; Catalog#11858-H08B; NP_000579.2; Met1-Phe152). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8484