Learn More
Aviva Systems Biology Corp IL3RA Antibody - C-terminal region (ARP63751_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63751_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL3RA. It was validated on Western Blot. . Alternate names: CD123, IL3R, IL3RAY, IL3RX, IL3RY, MGC34174, hIL-3RaApplicationWB. Species reactivity: Human
Catalog No. 50-288-073