Learn More
Sino Biological IL3RA Antibody (APC), Mouse MAb 100Tests
Supplier:  Sino Biological 10518MM57A100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL3RA (rh IL3RA; Catalog#10518-H08H; NP_002174.1; Met1-Arg305) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-224-6327