Learn More
Enzo Life Sciences IL-3 receptor alpha (human) monoclonal antibody (107D2.08) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2320100
Encompass_Preferred
Clone: 107D2.08. Isotype: IgG1. Immunogen: Sorted human tonsillar pDC. Species reactivity: Human Dog. Specificity: Recognies an epitope in the extracellular domain of CD123. Applications: Flow Cytometry, IHC (FS), IHC (PS), IP. Purity: Purified using QMA Hyper D ion exchange chromatography. Formulation: Liquid. In TRIS-NaCl, pH 8.
Catalog No. 50-201-2780