Learn More
Enzo Life Sciences IL-3 receptor alpha (human) monoclonal antibody (107D2.08) (Alexa Fluor 647) (100µg)
SDP
Catalog No. 502012778
Encompass_Preferred
Clone: 107D2.08. Isotype: IgG1. Immunogen: Sorted human tonsillar pDC. Species reactivity: Human Dog. Specificity: Recognies an epitope in the extracellular domain of CD123. Applications: Flow Cytometry. Purity: Purified using QMA Hyper D ion exchange chromatography. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-201-2778 Supplier Enzo Life Sciences Supplier No. ENZABS2300100
Clone: 107D2.08. Isotype: IgG1. Immunogen: Sorted human tonsillar pDC. Species reactivity: Human Dog. Specificity: Recognies an epitope in the extracellular domain of CD123. Applications: Flow Cytometry. Purity: Purified using QMA Hyper D ion exchange chromatography. Formulation: Liquid. In PBS containing 50% glycerol.