Learn More
Sino Biological Recombinant Human IL-3R alpha/CD123 Protein (hFc Tag)
Supplier:  Sino Biological 10518H02H50
Encompass_Preferred
A DNA sequence encoding the human IL3RA (NP_002174.1) (Met1-Arg305) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-4931