Learn More
Sino Biological Human IL3RA / CD123 Protein (His Tag) 1mg
Supplier:  Sino Biological 10518H08H1
Encompass_Preferred
A DNA sequence encoding the human IL3RA (NP_002174.1) extracellular domain (Met1-Arg305) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-211-6338