Learn More
Aviva Systems Biology Corp IL4 Antibody - middle region (ARP54330_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54330_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IL4. Alternate names: BSF1, IL-4, MGC79402, BCGF1, BSF-1, BCGF-1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61176