Learn More
ACROBiosystems Biotinylated Human Interleukin-4 Protein, Avitag™, His Tag, Lyophilized, 200 μg
Catalog No. 501658529
Encompass_Preferred
This biotinylated human interleukin-4 (IL-4) protein is expressed in human 293 (HEK293) cells and contains amino acids His 25 to Ser 153. As a critical cytokine, IL-4 induces the differentiation of naive helper T cells into Th2 cells and plays a significant role in mediating immune regulatory signals via STAT6. This product is engineered with an Avitag™ and a polyhistidine tag at the C-terminus, with biotinylation enzymatically performed on the single lysine residue within the Avitag™.
  • Greater than 95% purity as determined by SDS-PAGE
  • Calculated molecular weight of 17.6 kDa
  • Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4 with trehalose
  • Stable for 12 months at -20°C to -70°C in lyophilized state
Catalog No. 50-165-8529 Supplier ACROBiosystems Supplier No. IL4H82E0200UG
This biotinylated human interleukin-4 (IL-4) protein is expressed in human 293 (HEK293) cells and contains amino acids His 25 to Ser 153. As a critical cytokine, IL-4 induces the differentiation of naive helper T cells into Th2 cells and plays a significant role in mediating immune regulatory signals via STAT6. This product is engineered with an Avitag™ and a polyhistidine tag at the C-terminus, with biotinylation enzymatically performed on the single lysine residue within the Avitag™.
  • Greater than 95% purity as determined by SDS-PAGE
  • Calculated molecular weight of 17.6 kDa
  • Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4 with trehalose
  • Stable for 12 months at -20°C to -70°C in lyophilized state