Learn More
Sino Biological Human IL-4R / CD124 Protein (Fc Tag) 100ug
Supplier:  Sino Biological 10402H02H100
Encompass_Preferred
A DNA sequence encoding the human IL4R (NP_000409.1) (Met1-His232) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-4530