Learn More
ACROBiosystems Recombinant Protein;Cynomolgus / Rhesus macaque IL-4 R alpha / CD124 Protein, Fc Tag;HEK293;1MG;ILR,IL4R,IL-4R,IL4 R,CD124,IL-4RA,IL4RA,Interleukin-4 receptor,Interleukin 4 receptor,IL-4 R alpha
Catalog No. 501659109
Encompass_Preferred
Recombinant Protein;Cynomolgus / Rhesus macaque IL-4 R alpha / CD124 Protein, Fc Tag;1MG;Cynomolgus / Rhesus macaque IL-4 R alpha, Fc Tag (ILR-C5258) is expressed from human 293 cells (HEK293). It contains AA Met 26 - Arg 232 (Accession # G7Q0S7).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.4 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-9109 Supplier ACROBiosystems Supplier No. ILRC52581MG
Recombinant Protein;Cynomolgus / Rhesus macaque IL-4 R alpha / CD124 Protein, Fc Tag;1MG;Cynomolgus / Rhesus macaque IL-4 R alpha, Fc Tag (ILR-C5258) is expressed from human 293 cells (HEK293). It contains AA Met 26 - Arg 232 (Accession # G7Q0S7).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.4 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.